C12H316CE653 Chemcia Scientific, LLC

5-Chloromethyl-1H-pyrazole-4-carboxylic acid ethyl ester

  • MDL number MFCD03411879
  • Molecular Weight : 188.61498

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2176

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2176
Availability details for
Quantity
  • Availability
  • Add to Cart