C12H316CE656 Chemcia Scientific, LLC

2-Hydroxy-4-iodo-benzaldehyde

  • MDL number MFCD08457196
  • Cas Number : 38170-02-4
  • Molecular Weight : 248.0211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1766

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1766
Availability details for
Quantity
  • Availability
  • Add to Cart