C12H316CE657 Chemcia Scientific, LLC

8-Iodo-6-nitro-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • Cas Number : 1951441-31-8
  • Molecular Weight : 442.00933

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HBr
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1776

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1776
Availability details for
Quantity
  • Availability
  • Add to Cart