C12H316CE65E Chemcia Scientific, LLC

2-Iodo-6-methoxy-benzonitrile

  • MDL number MFCD18905184
  • Cas Number : 66195-38-8
  • Molecular Weight : 259.04752

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1780

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1780
Availability details for
Quantity
  • Availability
  • Add to Cart