C12H316CE66A Chemcia Scientific, LLC

(5-Bromo-3-hydroxymethyl-pyridin-2-yl)-methanol

  • MDL number MFCD22576225
  • Cas Number : 1356330-71-6
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromopyridine-2,3-diyl)dimethanol
Inchi Code 1S/C7H8BrNO2/c8-6-1-5(3-10)7(4-11)9-2-6/h1-2,10-11H,3-4H2
Inchi Key UIJUHQDWZRFCAZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1828

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1828
Availability details for
Quantity
  • Availability
  • Add to Cart