C12H316CE66B Chemcia Scientific, LLC

1,2,3,4-Tetrahydro-quinolin-7-ylamine hydrochloride

  • MDL number MFCD26743698
  • Cas Number : 1309976-14-4
  • Molecular Weight : 184.67036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HCl
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1844

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1844
Availability details for
Quantity
  • Availability
  • Add to Cart