C12H316CE66C Chemcia Scientific, LLC

1-(8-Nitro-3,4-dihydro-2H-quinolin-1-yl)-ethanone

  • MDL number MFCD23115372
  • Cas Number : 1290608-24-0
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(8-nitro-3,4-dihydroquinolin-1(2H)-yl)ethan-1-one
Inchi Code 1S/C11H12N2O3/c1-8(14)12-7-3-5-9-4-2-6-10(11(9)12)13(15)16/h2,4,6H,3,5,7H2,1H3
Inchi Key RCVSCVLVIRONHE-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1845

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1845
Availability details for
Quantity
  • Availability
  • Add to Cart