C12H316CE671 Chemcia Scientific, LLC

(4-Benzyl-piperazin-1-yl)-acetic acid

  • MDL number MFCD05668053
  • Cas Number : 119929-87-2
  • Molecular Weight : 234.30061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1837

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1837
Availability details for
Quantity
  • Availability
  • Add to Cart