C12H316CE673 Chemcia Scientific, LLC

4-Hydroxyimino-1-methanesulfonyl-pyrrolidine-3-carboxylic acid ethyl ester

  • MDL number MFCD23115371
  • Cas Number : 1421312-65-3
  • Molecular Weight : 250.27518

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1836

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1836
Availability details for
Quantity
  • Availability
  • Add to Cart