C12H316CE674 Chemcia Scientific, LLC

8-Nitro-1,2,3,4-tetrahydro-quinoline

  • MDL number MFCD00541473
  • Cas Number : 39217-93-1
  • Molecular Weight : 178.19225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1839

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1839
Availability details for
Quantity
  • Availability
  • Add to Cart