C12H316CE67E Chemcia Scientific, LLC

1-(6-Nitro-3,4-dihydro-2H-quinolin-1-yl)-ethanone

  • MDL number MFCD01463983
  • Cas Number : 14026-46-1
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-acetyl-6-nitro-1,2,3,4-tetrahydroquinoline
Inchi Code 1S/C11H12N2O3/c1-8(14)12-6-2-3-9-7-10(13(15)16)4-5-11(9)12/h4-5,7H,2-3,6H2,1H3
Inchi Key XXIBEYUXYNJBKQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1864

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1864
Availability details for
Quantity
  • Availability
  • Add to Cart