C12H316CE683 Chemcia Scientific, LLC

2-Iodo-4-methyl-6-nitro-phenylamine

  • MDL number MFCD23115398
  • Cas Number : 123158-77-0
  • Molecular Weight : 278.05044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1868

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1868
Availability details for
Quantity
  • Availability
  • Add to Cart