C12H316CE686 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-1H-indazol-5-ylamine hydrochloride

  • MDL number MFCD18838963
  • Cas Number : 1306606-59-6
  • Molecular Weight : 173.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5,6,7-tetrahydro-1H-indazol-5-amine hydrochloride
Inchi Code 1S/C7H11N3.ClH/c8-6-1-2-7-5(3-6)4-9-10-7;/h4,6H,1-3,8H2,(H,9,10);1H
Inchi Key SMQZJERKLBLABI-UHFFFAOYSA-N

Description

Purity 95%
Salt Data HCl
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1870

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1870
Availability details for
Quantity
  • Availability
  • Add to Cart