C12H316CE68A Chemcia Scientific, LLC

5-Amino-4-chloro-2-fluoro-benzoic acid methyl ester

  • MDL number MFCD17276528
  • Cas Number : 716362-14-0
  • Molecular Weight : 203.60189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1882

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1882
Availability details for
Quantity
  • Availability
  • Add to Cart