C12H316CE68C Chemcia Scientific, LLC

5-Bromo-2H-pyrazolo[3,4-b]pyridin-3-ylamine

  • MDL number MFCD22123459
  • Cas Number : 405224-24-0
  • Molecular Weight : 213.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2H-pyrazolo[3,4-b]pyridin-3-amine
Inchi Code 1S/C6H5BrN4/c7-3-1-4-5(8)10-11-6(4)9-2-3/h1-2H,(H3,8,9,10,11)
Inchi Key SSNUTEUZXZIYTB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2005

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2005
Availability details for
Quantity
  • Availability
  • Add to Cart