C12H316CE691 Chemcia Scientific, LLC

2-Chloro-6-methoxy-benzamide

  • MDL number MFCD11846167
  • Cas Number : 107485-43-8
  • Molecular Weight : 185.61146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1917

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1917
Availability details for
Quantity
  • Availability
  • Add to Cart