C12H316CE692 Chemcia Scientific, LLC

3-(4-Fluoro-phenyl)-6a-pyrrolidin-1-yl-3a,4,6,6a-tetrahydro-pyrrolo[3,4-d]isoxazole-5-carboxylic acid tert-butyl ester

  • MDL number MFCD23115399
  • Cas Number : 1421311-99-0
  • Molecular Weight : 375.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(4-fluorophenyl)-6a-(pyrrolidin-1-yl)-2,4,6,6a-tetrahydro-5H-pyrrolo[3,4-d]isoxazole-5-carboxylate
Inchi Code 1S/C20H26FN3O3/c1-19(2,3)26-18(25)23-12-16-17(14-6-8-15(21)9-7-14)22-27-20(16,13-23)24-10-4-5-11-24/h6-9,22H,4-5,10-13H2,1-3H3
Inchi Key ZBKQLZTULRZKSH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1876

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1876
Availability details for
Quantity
  • Availability
  • Add to Cart