C12H316CE693 Chemcia Scientific, LLC

3-(4-Fluoro-phenyl)-6a-pyrrolidin-1-yl-2,5,6,6a-tetrahydro-1-oxa-2,4-diaza-pentalene-4-carboxylic acid tert-butyl ester

  • MDL number MFCD23115400
  • Cas Number : 1421312-03-9
  • Molecular Weight : 375.44692

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1877

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1877
Availability details for
Quantity
  • Availability
  • Add to Cart