C12H316CE699 Chemcia Scientific, LLC

4-Formylamino-3-methoxy-benzoic acid methyl ester

  • MDL number MFCD09991719
  • Cas Number : 700834-18-0
  • Molecular Weight : 209.20347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1892

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1892
Availability details for
Quantity
  • Availability
  • Add to Cart