C12H316CE69A Chemcia Scientific, LLC

3-Chloro-5,7-dihydro-pyrrolo[3,4-c]pyridazine-6-carboxylic acid benzyl ester

  • MDL number MFCD23115402
  • Cas Number : 1421312-08-4
  • Molecular Weight : 289.72364

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1893

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1893
Availability details for
Quantity
  • Availability
  • Add to Cart