C12H316CE69C Chemcia Scientific, LLC

4-Methoxymethoxy-2-nitro-phenylamine

  • MDL number MFCD23115403
  • Cas Number : 54029-61-7
  • Molecular Weight : 198.1799

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1895

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1895
Availability details for
Quantity
  • Availability
  • Add to Cart