C12H316CE69E Chemcia Scientific, LLC

2-Chloromethyl-thieno[3,2-d]pyrimidin-4-ol

  • MDL number MFCD18162187
  • Cas Number : 147005-91-2
  • Molecular Weight : 200.6477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1897

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1897
Availability details for
Quantity
  • Availability
  • Add to Cart