C12H316CE6A3 Chemcia Scientific, LLC

3-Amino-5-nitro-benzo[b]thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD00245562
  • Cas Number : 34674-75-4
  • Molecular Weight : 252.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-5-nitro-1-benzothiophene-2-carboxylate
Inchi Code 1S/C10H8N2O4S/c1-16-10(13)9-8(11)6-4-5(12(14)15)2-3-7(6)17-9/h2-4H,11H2,1H3
Inchi Key WMZGLTDTPSMOMC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1900

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1900
Availability details for
Quantity
  • Availability
  • Add to Cart