C12H316CE6A4 Chemcia Scientific, LLC

2,5-Dichloro-pyridine 1-oxide

  • MDL number MFCD23115405
  • Cas Number : 53976-62-8
  • Molecular Weight : 163.99176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1902

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1902
Availability details for
Quantity
  • Availability
  • Add to Cart