C12H316CE6A7 Chemcia Scientific, LLC

(2-Phenyl-1H-imidazol-4-yl)-methanol

  • MDL number MFCD06202903
  • Cas Number : 43002-54-6
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-phenyl-1H-imidazol-4-yl)methanol
Inchi Code 1S/C10H10N2O/c13-7-9-6-11-10(12-9)8-4-2-1-3-5-8/h1-6,13H,7H2,(H,11,12)
Inchi Key GGRBEFVMJHQWFG-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1910

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1910
Availability details for
Quantity
  • Availability
  • Add to Cart