C12H316CE6A9 Chemcia Scientific, LLC

5-Trifluoromethyl-1H-benzoimidazole

  • MDL number MFCD06659629
  • Cas Number : 326-55-6
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(trifluoromethyl)-1H-benzimidazole
Inchi Code 1S/C8H5F3N2/c9-8(10,11)5-1-2-6-7(3-5)13-4-12-6/h1-4H,(H,12,13)
Inchi Key HUCHIALSXSAECU-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1905

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1905
Availability details for
Quantity
  • Availability
  • Add to Cart