C12H316CE6AB Chemcia Scientific, LLC

3,4-Diamino-phenol

  • MDL number MFCD06799539
  • Cas Number : 615-72-5
  • Molecular Weight : 124.14346

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1907

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1907
Availability details for
Quantity
  • Availability
  • Add to Cart