C12H316CE6AE Chemcia Scientific, LLC

2-Amino-3-bromo-6-methoxy-benzonitrile

  • MDL number MFCD24283992
  • Cas Number : 1440526-35-1
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-3-bromo-6-methoxybenzonitrile
Inchi Code 1S/C8H7BrN2O/c1-12-7-3-2-6(9)8(11)5(7)4-10/h2-3H,11H2,1H3
Inchi Key UHNXSVCUCBTPQT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1927

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1927
Availability details for
Quantity
  • Availability
  • Add to Cart