C12H316CE6AF Chemcia Scientific, LLC

6-Amino-3-bromo-2-methoxy-benzoic acid

  • MDL number MFCD23115401
  • Cas Number : 1421311-95-6
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-3-bromo-2-methoxybenzoic acid
Inchi Code 1S/C8H8BrNO3/c1-13-7-4(9)2-3-5(10)6(7)8(11)12/h2-3H,10H2,1H3,(H,11,12)
Inchi Key DZNJZWZQCNFPPK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1880

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1880
Availability details for
Quantity
  • Availability
  • Add to Cart