C12H316CE6B0 Chemcia Scientific, LLC

2-Amino-3-bromo-6-methoxy-benzoic acid

  • MDL number MFCD16743736
  • Cas Number : 1240480-64-1
  • Molecular Weight : 246.06186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1881

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1881
Availability details for
Quantity
  • Availability
  • Add to Cart