C12H316CE6B5 Chemcia Scientific, LLC

2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyridine-3,6-dicarboxylic acid 6-benzyl ester 3-methyl ester

  • MDL number MFCD23115407
  • Cas Number : 1421312-16-4
  • Molecular Weight : 346.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-benzyl 3-methyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Inchi Code 1S/C17H18N2O4S/c1-22-16(20)14-12-7-8-19(9-13(12)24-15(14)18)17(21)23-10-11-5-3-2-4-6-11/h2-6H,7-10,18H2,1H3
Inchi Key XFQBSOVIFCAAAN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1930

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1930
Availability details for
Quantity
  • Availability
  • Add to Cart