C12H316CE6B9 Chemcia Scientific, LLC

2-Hydrazino-nicotinonitrile

  • MDL number MFCD03086231
  • Cas Number : 368869-92-5
  • Molecular Weight : 134.14152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1925

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1925
Availability details for
Quantity
  • Availability
  • Add to Cart