C12H316CE6BB Chemcia Scientific, LLC

5-Nitro-2-oxo-1,2-dihydro-pyridine-3-carbonitrile

  • MDL number MFCD07065511
  • Cas Number : 31309-38-3
  • Molecular Weight : 165.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydroxy-5-nitronicotinonitrile
Inchi Code 1S/C6H3N3O3/c7-2-4-1-5(9(11)12)3-8-6(4)10/h1,3H,(H,8,10)
Inchi Key HLQZJYLEGMYYBY-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1920

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1920
Availability details for
Quantity
  • Availability
  • Add to Cart