C12H316CE6BE Chemcia Scientific, LLC

2-Oxo-6-trifluoromethyl-1,2-dihydro-pyridine-3-carbonitrile

  • MDL number MFCD09749653
  • Cas Number : 116548-04-0
  • Molecular Weight : 188.10996

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1922

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1922
Availability details for
Quantity
  • Availability
  • Add to Cart