C12H316CE6C2 Chemcia Scientific, LLC

2-Amino-7-oxo-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carbonitrile

  • MDL number MFCD01858636
  • Cas Number : 98899-30-0
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-7-oxo-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Inchi Code 1S/C9H8N2OS/c10-4-6-5-2-1-3-7(12)8(5)13-9(6)11/h1-3,11H2
Inchi Key XMZNDMVUUNFZEB-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1932

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1932
Availability details for
Quantity
  • Availability
  • Add to Cart