C12H316CE6CD Chemcia Scientific, LLC

2-Fluoro-3-iodo-benzonitrile

  • MDL number MFCD18396938
  • Cas Number : 211943-27-0
  • Molecular Weight : 247.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-3-iodobenzonitrile
Inchi Code 1S/C7H3FIN/c8-7-5(4-10)2-1-3-6(7)9/h1-3H
Inchi Key AFNXALOPDDVLMA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1941

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1941
Availability details for
Quantity
  • Availability
  • Add to Cart