C12H316CE6D0 Chemcia Scientific, LLC

2-Methylamino-benzonitrile

  • MDL number MFCD11155692
  • Cas Number : 17583-40-3
  • Molecular Weight : 132.16636

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1947

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1947
Availability details for
Quantity
  • Availability
  • Add to Cart