C12H316CE6D4 Chemcia Scientific, LLC

2,5-Dichloro-4-nitro-pyridine 1-oxide

  • MDL number MFCD11977416
  • Cas Number : 405230-81-1
  • Molecular Weight : 208.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dichloro-4-nitropyridine 1-oxide
Inchi Code 1S/C5H2Cl2N2O3/c6-3-2-8(10)5(7)1-4(3)9(11)12/h1-2H
Inchi Key XXLFCBBWDXLUOK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1952

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1952
Availability details for
Quantity
  • Availability
  • Add to Cart