C12H316CE6DB Chemcia Scientific, LLC

8-Bromo-6-fluoro-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD23115411
  • Cas Number : 1421312-07-3
  • Molecular Weight : 259.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-6-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C8H4BrFN2O2/c9-5-1-4(10)2-12-3-6(8(13)14)11-7(5)12/h1-3H,(H,13,14)
Inchi Key LBFJRGWQGMBRCK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Green Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1958

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1958
Availability details for
Quantity
  • Availability
  • Add to Cart