C12H316CE6DC Chemcia Scientific, LLC

6-Iodo-7-methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD23115412
  • Cas Number : 1036991-04-4
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-iodo-7-methylimidazo[1,2-a]pyridine
Inchi Code 1S/C8H7IN2/c1-6-4-8-10-2-3-11(8)5-7(6)9/h2-5H,1H3
Inchi Key YMOGVXPCGRRCAU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1959

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1959
Availability details for
Quantity
  • Availability
  • Add to Cart