C12H316CE6DE Chemcia Scientific, LLC

1-(4-Benzyl-piperazin-1-yl)-2-chloro-ethanone hydrochloride

  • MDL number MFCD01910940
  • Cas Number : 76087-89-3
  • Molecular Weight : 289.20721

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1961

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1961
Availability details for
Quantity
  • Availability
  • Add to Cart