C12H316CE6E1 Chemcia Scientific, LLC

2-Cyano-2-cyclopentylidene-acetamide

  • MDL number MFCD01923140
  • Cas Number : 875-61-6
  • Molecular Weight : 150.1817

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1965

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1965
Availability details for
Quantity
  • Availability
  • Add to Cart