C12H316CE6E6 Chemcia Scientific, LLC

7-Benzyl-2-methyl-5,6,7,8-tetrahydro-pyrido[3,4-d]pyrimidin-4-ol

  • MDL number MFCD17480434
  • Cas Number : 62259-92-1
  • Molecular Weight : 255.32224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1976

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1976
Availability details for
Quantity
  • Availability
  • Add to Cart