C12H316CE6EE Chemcia Scientific, LLC

6-Iodo-7-methyl-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD24843100
  • Cas Number : 1440526-41-9
  • Molecular Weight : 302.07274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1987

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1987
Availability details for
Quantity
  • Availability
  • Add to Cart