C12H316CE6F2 Chemcia Scientific, LLC

4-Chloro-2-methylamino-benzoic acid

  • MDL number MFCD17290555
  • Cas Number : 54675-18-2
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-(methylamino)benzoic acid
Inchi Code 1S/C8H8ClNO2/c1-10-7-4-5(9)2-3-6(7)8(11)12/h2-4,10H,1H3,(H,11,12)
Inchi Key AIKBBXVMNYCRDO-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1990

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1990
Availability details for
Quantity
  • Availability
  • Add to Cart