C12H316CE6F5 Chemcia Scientific, LLC

4-Amino-3-iodo-benzoic acid ethyl ester

  • MDL number MFCD00031420
  • Cas Number : 62875-84-7
  • Molecular Weight : 291.08995

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1998

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1998
Availability details for
Quantity
  • Availability
  • Add to Cart