C12H316CE6F6 Chemcia Scientific, LLC

4-Amino-3-iodo-benzoic acid

  • MDL number MFCD01660550
  • Cas Number : 2122-63-6
  • Molecular Weight : 263.03577

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1993

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1993
Availability details for
Quantity
  • Availability
  • Add to Cart