C12H316CE6F9 Chemcia Scientific, LLC

(4-Methoxy-3-nitro-phenyl)-acetonitrile

  • MDL number MFCD11118823
  • Cas Number : 56245-07-9
  • Molecular Weight : 192.17571

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1995

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1995
Availability details for
Quantity
  • Availability
  • Add to Cart