C12H316CE6FC Chemcia Scientific, LLC

4-Chloro-2-fluoro-5-iodo-benzoic acid

  • MDL number MFCD24107205
  • Cas Number : 1440526-33-9
  • Molecular Weight : 300.45656

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1981

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1981
Availability details for
Quantity
  • Availability
  • Add to Cart