C12H316CE6FD Chemcia Scientific, LLC

4-Chloro-5-chlorosulfonyl-2-hydroxy-benzoic acid

  • MDL number MFCD09044384
  • Cas Number : 14665-31-7
  • Molecular Weight : 271.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5-(chlorosulfonyl)-2-hydroxybenzoic acid
Inchi Code 1S/C7H4Cl2O5S/c8-4-2-5(10)3(7(11)12)1-6(4)15(9,13)14/h1-2,10H,(H,11,12)
Inchi Key CRNODOIWKMVMLY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1983

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1983
Availability details for
Quantity
  • Availability
  • Add to Cart